Monoolein 50%

Supplier: Ambeed
Warning

A570850-100G A570850-25G A570850-500G
76726-654EA 9 USD
76726-654 77602-402 77602-404
Monoolein 50%
Monoolein

Precaución: for Research Use Only

Formula: C₂₁H₄₀O₄
MDL Number: MFCD00042735
CAS Number: 111-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR