Nafamostat mesylate 98%

Supplier: Ambeed
Warning

A570309-100MG A570309-10MG A570309-1G A570309-1MG A570309-250MG A570309-50MG A570309-5G A570309-5MG
76726-638EA 46 USD
76726-638 76726-640 76726-642 76726-644 76726-646 76726-648 76726-650 76726-652
Nafamostat mesylate 98%
Nafamostat mesylate

Precaución: for Research Use Only

Formula: C₁₉H₁₇N₅O₂
MDL Number: MFCD00941430
CAS Number: 82956-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR