3-Pyrid-3-ylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Pyridin-3-yl-benzoic acid

A568539-1G A568539-250MG
76726-614EA 44.56 USD
76726-614 76726-616
3-Pyrid-3-ylbenzoic acid 97%
3-Pyrid-3-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD03426512
CAS Number: 4385-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR