N,N'-Bis(salicylidene)-1,2-propanediamine 97%

Supplier: Ambeed
Warning

Synonyms: N,N'-Bis(2-hydroxybenzylidene)-1,2-propanediamine, N,N'-Disalicylal-1,2-propanediamine

A566905-100G A566905-10G A566905-1G A566905-25G A566905-5G
76726-604EA 342.68 USD
76726-604 76726-606 76726-608 76726-610 76726-612
N,N'-Bis(salicylidene)-1,2-propanediamine 97%
N,N'-Bis(salicylidene)-1,2-propanediamine

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₂
MW: 282.34 g/mol
MDL Number: MFCD00020104
CAS Number: 94-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR