N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide 92%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pigment Red 179
A566091-100G
A566091-100MG
A566091-250MG
A566091-25G
A566091-5G
A566091-1G
76726-584EA
613.21
USD
76726-584
76726-586
76726-588
76726-590
76726-592
77669-496
N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide 92%
N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide
Precaución: for Research Use Only
Formula:
C₂₆H₁₄N₂O₄ MW: 418.41 g/mol |
MDL Number:
MFCD00071975 CAS Number: 5521-31-3 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...