Sodium (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoate 99%

Supplier: Ambeed
Warning

A565573-100G A565573-25G
76726-576EA 12.74 USD
76726-576 76726-578
Sodium (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoate 99%
Sodium (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NaO₁₂
MW: 380.27 g/mol
MDL Number: MFCD06658158
CAS Number: 27297-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR