Methyl-3-nitrosalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-3-nitrobenzoate

A565157-100G A565157-10G A565157-1G A565157-25G A565157-5G
76726-566EA 525.15 USD
76726-566 76726-568 76726-570 76726-572 76726-574
Methyl-3-nitrosalicylate 98%
Methyl-3-nitrosalicylate

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD00272263
CAS Number: 22621-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR