Phenol red sodium salt visual transition interval 6.8 - 8.2, yellow to red
Supplier: Ambeed
|
Warning
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
A563926-100G
A563926-25G
A563926-5G
A563926-500G
76726-552EA
97
USD
76726-552
76726-554
76726-556
77880-213
Phenol red sodium salt visual transition interval 6.8 - 8.2, yellow to red
Phenol red sodium salt
Precaución: for Research Use Only
Formula:
C₁₉H₁₃NaO₅S |
MDL Number:
MFCD00066901 CAS Number: 34487-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...