Oleic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: cis-9-Octadecenoic Anhydride, (9Z)-9-Octadecenoic acid anhydride

A563259-1G A563259-25G A563259-5G
76726-550EA 47.75 USD
76726-550 77669-492 77669-494
Oleic anhydride 98%
Oleic anhydride

Precaución: for Research Use Only

Formula: C₃₆H₆₆O₃
MW: 546.92 g/mol
MDL Number: MFCD00010459
CAS Number: 24909-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR