Phenyl bromoacetate 97%

Supplier: Ambeed
Warning

Synonyms: Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate

A561131-100G A561131-10G A561131-5G A561131-25G
76726-544EA 61.54 USD
76726-544 76726-546 76726-548 76843-976
Phenyl bromoacetate 97%
Phenyl bromoacetate

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD00192391
CAS Number: 620-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR