Rhodium(II) hexanoate, dimer 98%

Supplier: Ambeed
Warning

A561060-100MG A561060-250MG A561060-50MG
76726-532EA 173.99 USD
76726-532 76726-534 76726-536
Rhodium(II) hexanoate, dimer 98%
Rhodium(II) hexanoate, dimer

Precaución: for Research Use Only

MDL Number: MFCD00064723
CAS Number: 62728-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR