2',4'-Diethoxyacetophenone 97%

Supplier: Ambeed
Warning

A560235-1G A560235-250MG A560235-5G
76726-526EA 75.33 USD
76726-526 76726-528 76726-530
2',4'-Diethoxyacetophenone 97%
2',4'-Diethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.2536 g/mol
MDL Number: MFCD00238582
CAS Number: 22924-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR