2,9-Diphenylanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 97%

Supplier: Ambeed
Warning

A559086-1G A559086-250MG A559086-5G
76726-512EA 55 USD
76726-512 76726-514 76726-516
2,9-Diphenylanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 97%
2,9-Diphenylanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

Precaución: for Research Use Only

Formula: C₃₆H₁₈N₂O₄
MDL Number: MFCD00151605
CAS Number: 128-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR