N-Methyldicyclohexylamine 98%

Supplier: Ambeed
Danger

A559062-100G A559062-500G A559062-25G
76726-508EA 24 USD
76726-508 76726-510 77307-808
N-Methyldicyclohexylamine 98%
N-Methyldicyclohexylamine

Precaución: for Research Use Only

Formula: C₁₃H₂₅N
MDL Number: MFCD00014289
CAS Number: 7560-83-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR