N,N'-Bis(salicylidene)-1,3-propanediamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-((Propane-1,3-diylbis(azanylylidene))bis(methanylylidene))diphenol, N,N'-Disalicylal-1,3-propanediamine, N,N'-Bis(2-hydroxybenzylidene)-1,3-propanediamine

A558174-100G A558174-25G A558174-5G
76726-500EA 50.93 USD
76726-500 76726-502 76726-504
N,N'-Bis(salicylidene)-1,3-propanediamine 97%
N,N'-Bis(salicylidene)-1,3-propanediamine

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₂
MW: 282.34 g/mol
MDL Number: MFCD00002245
CAS Number: 120-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR