Methyl-5-allyl-3-methoxysalicylate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Allyl-3-methoxysalicylic Acid Methyl Ester, Methyl 5-Allyl-2-hydroxy-3-methoxybenzoate, Methyl 5-allyl-2-hydroxy-3-methoxybenzoate, 5-Allyl-2-hydroxy-3-methoxybenzoic Acid Methyl Ester

A557496-100G A557496-10G A557496-25G
76726-494EA 130 USD
76726-494 76726-496 76726-498
Methyl-5-allyl-3-methoxysalicylate 98%
Methyl-5-allyl-3-methoxysalicylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD04038819
CAS Number: 85614-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR