Phenyl benzoate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzoic acid phenyl ester
A556947-100G
A556947-25G
A556947-5G
A556947-500G
76726-486EA
56
USD
76726-486
76726-488
76726-490
77880-198
Phenyl benzoate 99%
Phenyl benzoate
Precaución: for Research Use Only
Formula:
C₁₃H₁₀O₂ |
MDL Number:
MFCD00003072 CAS Number: 93-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...