Sulfalene 99+%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulphonamide, N-(3-Methoxypyrazin-2-yl)sulphanilamide

A553516-100G A553516-1G A553516-5G A553516-25G
76726-472EA 221.73 USD
76726-472 76726-474 76726-476 77669-442
Sulfalene 99+%
Sulfalene

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₄O₃S
MW: 280.30 g/mol
MDL Number: MFCD00437754
CAS Number: 152-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR