m-Toluamide 99%

Supplier: Ambeed
Warning

Synonyms: 3-Methylbenzamide

A549643-100G A549643-25G
76726-458EA 108.22 USD
76726-458 76726-460
m-Toluamide 99%
m-Toluamide

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00007990
CAS Number: 618-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR