Tripotassium orthophosphate trihydrate 99%

Supplier: Ambeed
Warning

Synonyms: Potassium phosphate tribasic trihydrate, Potassium phosphate trihydrate

A548703-1000G A548703-100G A548703-25G A548703-500G
76726-450EA 15.92 USD
76726-450 76726-452 76726-454 76726-456
Tripotassium orthophosphate trihydrate 99%
Tripotassium orthophosphate trihydrate

Precaución: for Research Use Only

Formula: K₃O₄P·3H₂O
MW: 266.31 g/mol
MDL Number: MFCD00151709
CAS Number: 22763-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR