5-Methoxy-DL-tryptophan 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Trp(5-OMe)-OH, 2-Amino-3-(5-methoxy-1H-indol-3-yl)propanoic acid

A542738-1G A542738-250MG A542738-100MG
76726-386EA 221 USD
76726-386 77602-304 77669-394
5-Methoxy-DL-tryptophan 98%
5-Methoxy-DL-tryptophan

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃
MW: 234.255 g/mol
MDL Number: MFCD00005650
CAS Number: 28052-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR