Methyl-3-cyano-4-methoxybenzoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 3-cyano-4-methoxybenzoate

A542291-1G A542291-250MG A542291-5G
76726-380EA 20 USD
76726-380 76726-382 76726-384
Methyl-3-cyano-4-methoxybenzoate 98%
Methyl 3-cyano-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD00052930
CAS Number: 25978-74-9
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR