N-(2,5-Dimethoxyphenyl)benzamide 95+%

Supplier: Ambeed
Warning

A541914-1G A541914-100G A541914-25G A541914-5G
76726-378EA 11 USD
76726-378 77880-174 77880-175 77880-176
N-(2,5-Dimethoxyphenyl)benzamide 95+%
N-(2,5-Dimethoxyphenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₃
MDL Number: MFCD00035806
CAS Number: 135-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR