Methyldiphenylphosphine oxide 98%

Supplier: Ambeed
Warning

A541164-100G A541164-25G A541164-5G
76726-364EA 131.56 USD
76726-364 76726-366 76726-368
Methyldiphenylphosphine oxide 98%
Methyldiphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₃H₁₃OP
MW: 216.22 g/mol
MDL Number: MFCD00002081
CAS Number: 2129-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR