(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)acetyl chloride

A540216-5G A540216-10G
76726-362EA 224.92 USD
76726-362 77602-300
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl chloride 97%
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₃
MW: 223.62 g/mol
MDL Number: MFCD00192872
CAS Number: 6780-38-7
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR