Monomyristin 98+%

Supplier: Ambeed
Warning

Synonyms: 1-Monomyristoyl Glycerol, Glycerol alpha-Monomyristate, 2,3-Dihydroxypropyl tetradecanoate

A538715-1G A538715-250MG A538715-5G
76726-340EA 82.76 USD
76726-340 76726-342 76726-344
Monomyristin 98+%
Monomyristin

Precaución: for Research Use Only

Formula: C₁₇H₃₄O₄
MW: 302.46 g/mol
MDL Number: MFCD00046760
CAS Number: 589-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR