Sodium glycolate 98%

Supplier: Ambeed
Warning

A537716-100G A537716-25G A537716-500G A537716-1000G
76726-336EA 27 USD
76726-336 76726-338 77669-380 77872-900
Sodium glycolate 98%
Sodium glycolate

Precaución: for Research Use Only

Formula: C₂H₃NaO₃
MDL Number: MFCD00065358
CAS Number: 2836-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR