2-Chloro-4-chloromethylpyridine 97%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-4-(chloromethyl)pyridine

A537446-5G A537446-1G
76726-334EA 24 USD
76726-334 77602-292
2-Chloro-4-chloromethylpyridine 97%
2-Chloro-4-chloromethylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD09991655
CAS Number: 101990-73-2
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR