2-(1H-Indol-5-yl)-N-methylethanesulfonamide 97%

Supplier: Ambeed
Warning

A535377-1G A535377-250MG
76726-324EA 240.83 USD
76726-324 76726-326
2-(1H-Indol-5-yl)-N-methylethanesulfonamide 97%
2-(1H-Indol-5-yl)-N-methylethanesulfonamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₂S
MW: 238.30 g/mol
MDL Number: MFCD08458722
CAS Number: 98623-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR