Oxazole 98%

Supplier: Ambeed
Danger

A535158-10G A535158-1G A535158-5G A535158-25G
76726-318EA 52 USD
76726-318 76726-320 76726-322 77594-654
Oxazole 98%
Oxazole

Precaución: for Research Use Only

Formula: C₃H₃NO
MDL Number: MFCD00009751
CAS Number: 288-42-6
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR