Naphthalene-1,4-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A534132-10G A534132-250MG A534132-5G A534132-1G
76726-312EA 999 USD
76726-312 76726-314 76726-316 77669-366
Naphthalene-1,4-dicarbaldehyde 98%
Naphthalene-1,4-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈O₂
MW: 184.19 g/mol
MDL Number: MFCD01861387
CAS Number: 38153-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR