Clofibrate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate

A533783-100G A533783-1G A533783-25G A533783-5G
76726-302EA 111.4 USD
76726-302 76726-304 76726-306 76726-308
Clofibrate 98%
Clofibrate

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₃
MW: 242.70 g/mol
MDL Number: MFCD00000615
CAS Number: 637-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR