Octylbenzene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyloctane

A531302-100G A531302-25G A531302-5G
76726-296EA 114.58 USD
76726-296 76726-298 76726-300
Octylbenzene 95%
Octylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₂
MW: 190.33 g/mol
MDL Number: MFCD00009564
CAS Number: 2189-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR