N,N-Dimethylbenzamide 95%

Supplier: Ambeed
Warning

A530130-100G A530130-1G A530130-25G A530130-5G
76726-282EA 71.09 USD
76726-282 76726-284 76726-286 76726-288
N,N-Dimethylbenzamide 95%
N,N-Dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD00008320
CAS Number: 611-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR