Methyl (6-chloro-1H-benzo[d]imidazol-2-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Fenbendazole Related Compound B

A529665-1G A529665-250MG A529665-100MG A529665-5G
76726-278EA 231 USD
76726-278 76726-280 77669-350 77880-149
Methyl (6-chloro-1H-benzo[d]imidazol-2-yl)carbamate 98%
Methyl (6-chloro-1H-benzo[d]imidazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₈ClN₃O₂
MW: 225.63 g/mol
MDL Number: MFCD08234600
CAS Number: 20367-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR