1-Benzoylimidazole 95%

Supplier: Ambeed
Warning

Synonyms: (1H-Imidazol-1-yl)(phenyl)methanone

A529538-1G A529538-250MG A529538-25G A529538-5G
76726-274EA 28.65 USD
76726-274 76726-276 77669-346 77669-348
1-Benzoylimidazole 95%
1-Benzoylimidazole

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.1833 g/mol
MDL Number: MFCD00014499
CAS Number: 10364-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR