Platinum(IV) chloride 97%

Supplier: Ambeed
Danger

A528414-100MG A528414-10G A528414-1G A528414-250MG A528414-5G A528414-25G
76726-254EA 6 USD
76726-254 76726-256 76726-258 76726-260 76726-262 77669-342
Platinum(IV) chloride 97%
Platinum(IV) chloride

Precaución: for Research Use Only

Formula: Cl₄Pt
MDL Number: MFCD00011182
CAS Number: 13454-96-1
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR