Tetradecylamine 99%

Supplier: Ambeed
Danger

Synonyms: 1-Tetradecylamine, 1-Aminotetradecane, n-Tetradecylamine, Tetradecan-1-amine

A527964-25G A527964-100G
76726-252EA 23 USD
76726-252 77669-340
Tetradecylamine 99%
Tetradecylamine

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₂CH₂NH₂
MW: 213.41 g/mol
MDL Number: MFCD00008157
CAS Number: 2016-42-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR