Phenyl methacrylate 97% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A525305-25G A525305-5G
76726-232EA 30 USD
76726-232 76726-234
Phenyl methacrylate 97% stabilized
Phenyl methacrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00048117
CAS Number: 2177-70-0
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR