N,N-Diisobutyl-2-(octyl(phenyl)phosphoryl)acetamide 95%

Supplier: Ambeed
Warning

A525255-5G A525255-1G A525255-250MG
76726-230EA 109 USD
76726-230 77572-674 77880-126
N,N-Diisobutyl-2-(octyl(phenyl)phosphoryl)acetamide 95%
N,N-Diisobutyl-2-(octyl(phenyl)phosphoryl)acetamide

Precaución: for Research Use Only

Formula: C₂₄H₄₂NO₂P
MDL Number: MFCD00079653
CAS Number: 83242-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR