2-(Benzylthio)nicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzylthio)pyridine-3-carboxylic Acid

A524524-1G A524524-250MG A524524-5G
76726-224EA 14 USD
76726-224 76726-226 76726-228
2-(Benzylthio)nicotinic acid 98%
2-(Benzylthio)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂S
MDL Number: MFCD00661380
CAS Number: 112811-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR