N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 50%

Supplier: Ambeed
Warning

Synonyms: N,N,N,N-Tetrakis(2-hydroxyethyl)ethylenediamine

A519492-100G A519492-25G A519492-500G A519492-5G
76726-198EA 11.67 USD
76726-198 76726-200 76726-202 76726-204
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 50%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₀H₂₄N₂O₄
MW: 236.3086 g/mol
MDL Number: MFCD00002854
CAS Number: 140-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR