Potassium trifluoro(pentyl)borate 97%
Supplier: Ambeed
|
Danger
|
A517851-1G
A517851-250MG
A517851-5G
76726-192EA
28
USD
76726-192
76726-194
76726-196
Potassium trifluoro(pentyl)borate 97%
Potassium trifluoro(pentyl)borate
Precaución: for Research Use Only
Formula:
C₅H₁₁BF₃K |
MDL Number:
MFCD07368249 CAS Number: 872054-60-9 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...