Potassium (2-methoxyphenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(2-methoxyphenyl)borate, (2-Methoxyphenyl)trifluoroboric acid potassium salt

A515509-10G A515509-1G A515509-250MG A515509-25G A515509-5G
76726-158EA 85 USD
76726-158 76726-160 76726-162 76726-164 76726-166
Potassium (2-methoxyphenyl)trifluoroborate 98%
Potassium (2-methoxyphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
MDL Number: MFCD04115756
CAS Number: 236388-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR