Platinum disulfide 98%

Supplier: Ambeed
Warning

A514677-100MG A514677-1G A514677-250MG A514677-5G
76726-152EA 53 USD
76726-152 76726-154 76726-156 77880-111
Platinum disulfide 98%
Platinum disulphide

Precaución: for Research Use Only

Formula: PtS₂
MDL Number: MFCD00016293
CAS Number: 12038-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR