o-Tolylacetonitrile 98%

Supplier: Ambeed
Warning

A512700-1000G A512700-100G A512700-10G A512700-25G A512700-500G
76726-142EA 142 USD
76726-142 76726-144 76726-146 76726-148 76726-150
o-Tolylacetonitrile 98%
o-Tolylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₉N
MDL Number: MFCD00001904
CAS Number: 22364-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR