4-Aminophenyl α-D-mannopyranoside 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3S,4S,5S,6R)-2-(4-Aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A512234-100MG A512234-250MG A512234-50MG
76726-136EA 156 USD
76726-136 76726-138 76726-140
4-Aminophenyl α-D-mannopyranoside 98%
4-Aminophenyl α-D-mannopyranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₆
MW: 271.266 g/mol
MDL Number: MFCD00067366
CAS Number: 34213-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR