Sodium tetrakis(4-fluorophenyl)borate 98%

Supplier: Ambeed
Warning

A511969-100MG A511969-1G A511969-250MG A511969-5G
76726-128EA 48.81 USD
76726-128 76726-130 76726-132 76726-134
Sodium tetrakis(4-fluorophenyl)borate 98%
Sodium tetrakis(4-fluorophenyl)borate

Precaución: for Research Use Only

Formula: C₂₄H₁₆BF₄Na
MW: 414.17 g/mol
MDL Number: MFCD00010732
CAS Number: 25776-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR