SATA (N-Succinimidyl S-Acetylthioglycolate) 98%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

A508892-250MG A508892-5G A508892-1G A508892-100MG
76726-120EA 152 USD
76726-120 76726-122 77568-992 77602-172
SATA (N-Succinimidyl S-Acetylthioglycolate) 98%
SATA (N-Succinimidyl S-Acetylthioglycolate)

Precaución: For research use only.

Formula: C₈H₉NO₅S
MW: 231.22 g/mol
MDL Number: MFCD00036891
CAS Number: 76931-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR