Pyrazine 98%

Supplier: Ambeed
Danger

A508019-10G A508019-25G A508019-5G
76726-104EA 6.37 USD
76726-104 76726-106 76726-108
Pyrazine 98%
Pyrazine

Precaución: for Research Use Only

Formula: C₄H₄N₂
MW: 80.09 g/mol
MDL Number: MFCD00006122
CAS Number: 290-37-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR